Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608473 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANAPC1 - anaphase promoting complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022662.3 | Intron | NP_073153.1 | ||||
XM_011511633.2 | Intron | XP_011509935.1 | ||||
XM_011511634.2 | Intron | XP_011509936.1 | ||||
XM_011511636.2 | Intron | XP_011509938.1 | ||||
XM_011511637.2 | Intron | XP_011509939.1 | ||||
XM_017004710.1 | Intron | XP_016860199.1 | ||||
XM_017004711.1 | Intron | XP_016860200.1 | ||||
XM_017004712.1 | Intron | XP_016860201.1 | ||||
XM_017004713.1 | Intron | XP_016860202.1 | ||||
XM_017004714.1 | Intron | XP_016860203.1 | ||||
XM_017004715.1 | Intron | XP_016860204.1 |
Set Membership: |
HapMap |