Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAGGGACTAGTCGCAGCTTTC[G/A]TCGCCGCCGATTCGTCAAGGTCCCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609085 MIM: 610930 MIM: 611052 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXL19 - F-box and leucine rich repeat protein 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORAI3 - ORAI calcium release-activated calcium modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD1A - SET domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014712.2 | 447 | UTR 5 | NP_055527.1 | |||
XM_005255723.1 | 447 | Intron | XP_005255780.1 | |||
XM_006721106.3 | 447 | Intron | XP_006721169.1 | |||
XM_017023909.1 | 447 | UTR 5 | XP_016879398.1 |
Set Membership: |
HapMap |