Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCAGGAACAGAATGAGGGTTC[A/G]GGGGATGGGGCCTGGGGTGAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602989 MIM: 609973 MIM: 609712 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLK2 - CDC like kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HCN3 - hyperpolarization activated cyclic nucleotide gated potassium channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020897.2 | Intron | NP_065948.1 | ||||
XM_011509816.2 | Intron | XP_011508118.1 | ||||
XM_011509817.2 | Intron | XP_011508119.1 | ||||
XM_011509818.2 | Intron | XP_011508120.1 | ||||
XM_017001918.1 | Intron | XP_016857407.1 |
PKLR - pyruvate kinase, liver and RBC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |