Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123885 | ||||||||||||||||||||||||||||||||
Literature Links: |
S100A8 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
S100A8 - S100 calcium binding protein A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319196.1 | Intron | NP_001306125.1 | ||||
NM_001319197.1 | Intron | NP_001306126.1 | ||||
NM_001319198.1 | Intron | NP_001306127.1 | ||||
NM_001319201.1 | Intron | NP_001306130.1 | ||||
NM_002964.4 | Intron | NP_002955.2 |
Set Membership: |
HapMap |