Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
A1CF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
A1CF - APOBEC1 complementation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198818.1 | 2482 | UTR 3 | NP_001185747.1 | |||
NM_001198819.1 | 2482 | UTR 3 | NP_001185748.1 | |||
NM_001198820.1 | 2482 | UTR 3 | NP_001185749.1 | |||
NM_014576.3 | 2482 | UTR 3 | NP_055391.2 | |||
NM_138932.2 | 2482 | UTR 3 | NP_620310.1 | |||
NM_138933.2 | 2482 | UTR 3 | NP_620311.1 | |||
XM_005269718.2 | 2482 | UTR 3 | XP_005269775.1 | |||
XM_005269720.3 | 2482 | UTR 3 | XP_005269777.1 | |||
XM_011539729.2 | 2482 | UTR 3 | XP_011538031.1 | |||
XM_011539730.2 | 2482 | UTR 3 | XP_011538032.1 | |||
XM_017016160.1 | 2482 | UTR 3 | XP_016871649.1 |