Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 613450 | ||||||||||||||||||||||||||
Literature Links: |
MZT2B PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MZT2B - mitotic spindle organizing protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263795.4 | Intron | XP_005263852.1 | ||||
XM_011511886.2 | Intron | XP_011510188.1 | ||||
XM_011511887.2 | Intron | XP_011510189.1 | ||||
XM_011511888.2 | Intron | XP_011510190.1 | ||||
XM_011511889.2 | Intron | XP_011510191.1 | ||||
XM_011511890.2 | Intron | XP_011510192.1 |
TUBA3E - tubulin alpha 3e | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207312.2 | Intron | NP_997195.1 |
Set Membership: |
HapMap |