Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGATGGCCTCGATGAGGAAGCCCG[A/C]CTCCTCCTGGATGCGCTCCTCGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608054 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP2A7 - cytochrome P450 family 2 subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000764.2 | Intron | NP_000755.2 | ||||
NM_030589.2 | Intron | NP_085079.2 | ||||
XM_017026383.1 | Intron | XP_016881872.1 |
Set Membership: |
DME Validated Inventoried |