Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608469 MIM: 602721 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX17 - DEAD-box helicase 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DMC1 - DNA meiotic recombinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278208.1 | Intron | NP_001265137.1 | ||||
NM_007068.3 | Intron | NP_008999.2 | ||||
XM_006724112.2 | Intron | XP_006724175.1 | ||||
XM_011529834.1 | Intron | XP_011528136.1 | ||||
XM_011529835.2 | Intron | XP_011528137.1 | ||||
XM_011529837.1 | Intron | XP_011528139.1 | ||||
XM_011529838.1 | Intron | XP_011528140.1 | ||||
XM_017028542.1 | Intron | XP_016884031.1 |
Set Membership: |
HapMap |