Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTTCCCACTTATTGTGCCTCA[C/T]AATTTTATTTTTCTTCTTTCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603576 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTMR10 - myotubularin related protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPM1 - transient receptor potential cation channel subfamily M member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252020.1 | 5814 | UTR 3 | NP_001238949.1 | |||
NM_001252024.1 | 5814 | UTR 3 | NP_001238953.1 | |||
NM_001252030.1 | 5814 | Intron | NP_001238959.1 | |||
NM_002420.5 | 5814 | UTR 3 | NP_002411.3 |
Set Membership: |
HapMap |