Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATCTTGTTCACAGACGGCGACC[A/G]CGGCGACCCCCCTTCCTGCGAGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130620 | ||||||||||||||||||||
Literature Links: |
RPS14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPS14 - ribosomal protein S14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025070.1 | 714 | Silent Mutation | CGC,CGT | R,R 147 | NP_001020241.1 | |
NM_001025071.1 | 714 | Silent Mutation | CGC,CGT | R,R 147 | NP_001020242.1 | |
NM_005617.3 | 714 | Intron | NP_005608.1 |