Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC6A1 - solute carrier family 6 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003042.3 | Intron | NP_003033.3 | ||||
XM_005265410.4 | Intron | XP_005265467.1 | ||||
XM_005265411.4 | Intron | XP_005265468.1 | ||||
XM_006713306.3 | Intron | XP_006713369.1 | ||||
XM_011534025.2 | Intron | XP_011532327.1 | ||||
XM_011534027.2 | Intron | XP_011532329.1 | ||||
XM_017007071.1 | Intron | XP_016862560.1 | ||||
XM_017007072.1 | Intron | XP_016862561.1 |