Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607910 | ||||||||||||||||||||
Literature Links: |
KIF9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF9 - kinesin family member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLHL18 - kelch like family member 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025010.4 | Intron | NP_079286.2 | ||||
XM_005265001.3 | Intron | XP_005265058.2 | ||||
XM_005265002.4 | Intron | XP_005265059.2 | ||||
XM_005265003.3 | Intron | XP_005265060.2 | ||||
XM_006713079.1 | Intron | XP_006713142.1 | ||||
XM_017006033.1 | Intron | XP_016861522.1 | ||||
XM_017006034.1 | Intron | XP_016861523.1 | ||||
XM_017006035.1 | Intron | XP_016861524.1 |