Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606884 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBM5 - RNA binding motif protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005778.3 | Intron | NP_005769.1 | ||||
XM_006712917.1 | Intron | XP_006712980.1 | ||||
XM_011533261.1 | Intron | XP_011531563.1 | ||||
XM_011533262.1 | Intron | XP_011531564.1 | ||||
XM_017005503.1 | Intron | XP_016860992.1 | ||||
XM_017005504.1 | Intron | XP_016860993.1 | ||||
XM_017005505.1 | Intron | XP_016860994.1 | ||||
XM_017005506.1 | Intron | XP_016860995.1 | ||||
XM_017005507.1 | Intron | XP_016860996.1 |
RBM5-AS1 - RBM5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |