Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGACAAAGAAAGAGAGTAGTCC[A/G]TGAGAAAAGAAGAAAACCAAGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613528 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR302E PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR302E - microRNA 302e | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT9 - synaptotagmin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175733.3 | Intron | NP_783860.1 | ||||
XM_011519900.2 | Intron | XP_011518202.1 | ||||
XM_011519901.2 | Intron | XP_011518203.1 | ||||
XM_011519902.2 | Intron | XP_011518204.1 | ||||
XM_011519904.2 | Intron | XP_011518206.1 | ||||
XM_011519906.2 | Intron | XP_011518208.1 | ||||
XM_011519907.2 | Intron | XP_011518209.1 |
Set Membership: |
HapMap |