Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603265 MIM: 604161 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARID3A - AT-rich interaction domain 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005224.2 | Intron | NP_005215.1 | ||||
XM_005259513.4 | Intron | XP_005259570.1 | ||||
XM_005259514.4 | Intron | XP_005259571.1 | ||||
XM_017026445.1 | Intron | XP_016881934.1 | ||||
XM_017026446.1 | Intron | XP_016881935.1 | ||||
XM_017026447.1 | Intron | XP_016881936.1 |
KISS1R - KISS1 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |