Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCCTCTTCCAGCAGCTCCGAG[C/G]GCTCTCAGAGAAAAACGAAATTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601990 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TP73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TP73 - tumor protein p73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126240.2 | Intron | NP_001119712.1 | ||||
NM_001126241.2 | Intron | NP_001119713.1 | ||||
NM_001126242.2 | Intron | NP_001119714.1 | ||||
NM_001204184.1 | Intron | NP_001191113.1 | ||||
NM_001204185.1 | Intron | NP_001191114.1 | ||||
NM_001204186.1 | Intron | NP_001191115.1 | ||||
NM_001204187.1 | Intron | NP_001191116.1 | ||||
NM_001204188.1 | Intron | NP_001191117.1 | ||||
NM_001204189.1 | Intron | NP_001191118.1 | ||||
NM_001204190.1 | Intron | NP_001191119.1 | ||||
NM_001204191.1 | Intron | NP_001191120.1 | ||||
NM_001204192.1 | Intron | NP_001191121.1 | ||||
NM_005427.3 | Intron | NP_005418.1 |