Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACCCTCTTCCCTCTTCCCCATC[G/T]GAGAACAGGGTAGCCTTTAATGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPRX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPRX - divergent-paired related homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012728.1 | Intron | NP_001012746.1 | ||||
XM_011527011.2 | Intron | XP_011525313.1 | ||||
XM_011527012.2 | Intron | XP_011525314.1 |
LOC284379 - solute carrier family 7 member 3 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |