Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606898 MIM: 606900 | ||||||||||||||||||||
Literature Links: |
CACNG6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNG6 - calcium voltage-gated channel auxiliary subunit gamma 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031897.2 | Intron | NP_114103.2 | ||||
NM_145814.1 | Intron | NP_665813.1 | ||||
NM_145815.1 | Intron | NP_665814.1 |
CACNG8 - calcium voltage-gated channel auxiliary subunit gamma 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |