Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616378 MIM: 616097 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf98 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf98 - chromosome 11 open reading frame 98 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBHD1 - LBH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024099.3 | Intron | NP_077004.2 |
METTL12 - methyltransferase like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA57 - small nucleolar RNA, H/ACA box 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN1 - UBX domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UQCC3 - ubiquinol-cytochrome c reductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085372.2 | Intron | NP_001078841.1 |
Set Membership: |
HapMap |