Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGTTTGGCTTAGGCACACTTAC[C/T]GACTCCACAGTTGGAACAATGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF121 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300926.1 | Intron | NP_001287855.1 | ||||
NM_018320.4 | Intron | NP_060790.2 | ||||
XM_006718629.1 | Intron | XP_006718692.1 | ||||
XM_006718630.1 | Intron | XP_006718693.1 | ||||
XM_006718632.1 | Intron | XP_006718695.1 | ||||
XM_011545150.1 | Intron | XP_011543452.1 |
Set Membership: |
HapMap |