Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137207 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC32 - leucine rich repeat containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128922.1 | 2868 | Intron | NP_001122394.1 | |||
NM_005512.2 | 2868 | Intron | NP_005503.1 | |||
XM_005273902.3 | 2868 | UTR 3 | XP_005273959.1 | |||
XM_005273903.4 | 2868 | Intron | XP_005273960.2 | |||
XM_011544935.2 | 2868 | Intron | XP_011543237.1 | |||
XM_017017527.1 | 2868 | UTR 3 | XP_016873016.1 |
Set Membership: |
HapMap |