Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf100 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C1orf100 - chromosome 1 open reading frame 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012970.2 | Intron | NP_001012988.1 | ||||
NM_001276348.1 | Intron | NP_001263277.1 | ||||
NM_001276349.1 | Intron | NP_001263278.1 | ||||
XM_011544124.2 | Intron | XP_011542426.1 | ||||
XM_011544125.2 | Intron | XP_011542427.1 | ||||
XM_011544128.2 | Intron | XP_011542430.1 | ||||
XM_011544129.2 | Intron | XP_011542431.1 | ||||
XM_011544130.2 | Intron | XP_011542432.1 | ||||
XM_017000571.1 | Intron | XP_016856060.1 | ||||
XM_017000572.1 | Intron | XP_016856061.1 |