Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf100 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf100 - chromosome 1 open reading frame 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012970.2 | Intron | NP_001012988.1 | ||||
NM_001276348.1 | Intron | NP_001263277.1 | ||||
NM_001276349.1 | Intron | NP_001263278.1 | ||||
XM_011544124.2 | Intron | XP_011542426.1 | ||||
XM_011544125.2 | Intron | XP_011542427.1 | ||||
XM_011544128.2 | Intron | XP_011542430.1 | ||||
XM_011544129.2 | Intron | XP_011542431.1 | ||||
XM_011544130.2 | Intron | XP_011542432.1 | ||||
XM_017000571.1 | Intron | XP_016856060.1 | ||||
XM_017000572.1 | Intron | XP_016856061.1 |