Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGAAAGAAGCAATGTTCCATGG[C/T]CTGGAAAACAGTGGGTATTTATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606208 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928414 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928414 - uncharacterized LOC101928414 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC28A2 - solute carrier family 28 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004212.3 | Intron | NP_004203.2 | ||||
XM_011522198.2 | Intron | XP_011520500.1 | ||||
XM_011522199.1 | Intron | XP_011520501.1 | ||||
XM_011522200.2 | Intron | XP_011520502.1 | ||||
XM_011522201.2 | Intron | XP_011520503.1 | ||||
XM_011522202.2 | Intron | XP_011520504.1 |
Set Membership: |
HapMap |