Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PARP16 - poly(ADP-ribose) polymerase family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316943.1 | 1434 | Intron | NP_001303872.1 | |||
NM_001316944.1 | 1434 | Intron | NP_001303873.1 | |||
NM_017851.5 | 1434 | Intron | NP_060321.3 | |||
XM_006720589.2 | 1434 | UTR 3 | XP_006720652.1 | |||
XM_006720590.2 | 1434 | UTR 3 | XP_006720653.1 | |||
XM_006720592.2 | 1434 | Intron | XP_006720655.1 | |||
XM_011521742.2 | 1434 | Intron | XP_011520044.1 | |||
XM_017022386.1 | 1434 | Intron | XP_016877875.1 | |||
XM_017022387.1 | 1434 | Intron | XP_016877876.1 | |||
XM_017022388.1 | 1434 | Intron | XP_016877877.1 | |||
XM_017022389.1 | 1434 | Intron | XP_016877878.1 |