Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600700 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ42393 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLJ42393 - uncharacterized LOC401105 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LPP - LIM domain containing preferred translocation partner in lipoma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167671.2 | Intron | NP_001161143.1 | ||||
NM_001167672.2 | Intron | NP_001161144.1 | ||||
NM_005578.4 | Intron | NP_005569.1 | ||||
XM_005247446.4 | Intron | XP_005247503.1 | ||||
XM_005247450.4 | Intron | XP_005247507.1 | ||||
XM_005247451.4 | Intron | XP_005247508.1 | ||||
XM_005247453.2 | Intron | XP_005247510.1 | ||||
XM_011512820.2 | Intron | XP_011511122.1 | ||||
XM_011512823.2 | Intron | XP_011511125.1 | ||||
XM_011512827.2 | Intron | XP_011511129.1 | ||||
XM_011512828.2 | Intron | XP_011511130.1 | ||||
XM_011512831.2 | Intron | XP_011511133.1 | ||||
XM_011512833.2 | Intron | XP_011511135.1 | ||||
XM_011512834.2 | Intron | XP_011511136.1 | ||||
XM_011512836.2 | Intron | XP_011511138.1 | ||||
XM_017006377.1 | Intron | XP_016861866.1 | ||||
XM_017006378.1 | Intron | XP_016861867.1 | ||||
XM_017006379.1 | Intron | XP_016861868.1 | ||||
XM_017006380.1 | Intron | XP_016861869.1 | ||||
XM_017006381.1 | Intron | XP_016861870.1 | ||||
XM_017006382.1 | Intron | XP_016861871.1 |
Set Membership: |
HapMap |