Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAGTCCTGGCACACCCTTACAG[A/G]TGTTCACCCAAGTTTTAATCTATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603930 MIM: 608830 MIM: 612012 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPHN PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPHN - gephyrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RDH12 - retinol dehydrogenase 12 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFYVE26 - zinc finger FYVE-type containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015346.3 | Intron | NP_056161.2 | ||||
XM_011536609.2 | Intron | XP_011534911.1 | ||||
XM_017021124.1 | Intron | XP_016876613.1 | ||||
XM_017021125.1 | Intron | XP_016876614.1 | ||||
XM_017021126.1 | Intron | XP_016876615.1 | ||||
XM_017021127.1 | Intron | XP_016876616.1 | ||||
XM_017021128.1 | Intron | XP_016876617.1 |