Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607806 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTOP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTOP1 - otopetrin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM128 - transmembrane protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297551.1 | 838 | UTR 3 | NP_001284480.1 | |||
NM_001297552.1 | 838 | UTR 3 | NP_001284481.1 | |||
NM_032927.3 | 838 | UTR 3 | NP_116316.1 | |||
XM_005248034.2 | 838 | UTR 3 | XP_005248091.1 |