Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATACGAAAAATATGTTCTTTAAA[A/G]CCTAGGAAGCTTTAATACATTCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606821 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAP29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCAP29 - B-cell receptor-associated protein 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COG5 - component of oligomeric golgi complex 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUS4L - dihydrouridine synthase 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270419.1 | Intron | NP_001257348.1 | ||||
NM_181581.2 | Intron | NP_853559.1 | ||||
XM_005250125.4 | Intron | XP_005250182.1 | ||||
XM_006715844.2 | Intron | XP_006715907.1 | ||||
XM_006715845.3 | Intron | XP_006715908.1 | ||||
XM_011515766.2 | Intron | XP_011514068.1 | ||||
XM_011515767.2 | Intron | XP_011514069.1 | ||||
XM_017011708.1 | Intron | XP_016867197.1 | ||||
XM_017011709.1 | Intron | XP_016867198.1 | ||||
XM_017011710.1 | Intron | XP_016867199.1 | ||||
XM_017011711.1 | Intron | XP_016867200.1 | ||||
XM_017011712.1 | Intron | XP_016867201.1 | ||||
XM_017011713.1 | Intron | XP_016867202.1 |
Set Membership: |
HapMap |