Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF562 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF562 - zinc finger protein 562 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130031.1 | 1519 | Silent Mutation | CAA,CAG | Q,Q 389 | NP_001123503.1 | |
NM_001130032.1 | 1519 | Silent Mutation | CAA,CAG | Q,Q 389 | NP_001123504.1 | |
NM_001300885.1 | 1519 | Silent Mutation | CAA,CAG | Q,Q 388 | NP_001287814.1 | |
NM_017656.3 | 1519 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_060126.2 | |
XM_005259941.3 | 1519 | Silent Mutation | CAA,CAG | Q,Q 389 | XP_005259998.1 | |
XM_005259944.3 | 1519 | Silent Mutation | CAA,CAG | Q,Q 352 | XP_005260001.1 | |
XM_005259951.3 | 1519 | Silent Mutation | CAA,CAG | Q,Q 273 | XP_005260008.1 | |
XM_005259952.3 | 1519 | Silent Mutation | CAA,CAG | Q,Q 246 | XP_005260009.1 | |
XM_011528100.2 | 1519 | Silent Mutation | CAA,CAG | Q,Q 320 | XP_011526402.1 | |
XM_011528102.2 | 1519 | Silent Mutation | CAA,CAG | Q,Q 274 | XP_011526404.1 | |
XM_017026898.1 | 1519 | Silent Mutation | CAA,CAG | Q,Q 388 | XP_016882387.1 | |
XM_017026899.1 | 1519 | Silent Mutation | CAA,CAG | Q,Q 246 | XP_016882388.1 |