Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGACTACAAGATTCGAGTTACC[A/G]TGCCCGATTAATTTTTGTATTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606106 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC44A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC44A2 - solute carrier family 44 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145056.1 | Intron | NP_001138528.1 | ||||
NM_020428.3 | Intron | NP_065161.3 | ||||
XM_005259997.1 | Intron | XP_005260054.1 | ||||
XM_005259999.1 | Intron | XP_005260056.1 |
Set Membership: |
HapMap |