Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGTGTAGTTGATCTAGTTAAC[C/T]AGGTATCTGTCTTCAGGTTTCTTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604052 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TNFSF15 - tumor necrosis factor superfamily member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204344.1 | 2151 | UTR 3 | NP_001191273.1 | |||
NM_005118.3 | 2151 | UTR 3 | NP_005109.2 |