Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCATCCCGGCTGCCCATGCTC[C/T]GGAGAGAGGAGGTTTTCTTTTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUSD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUSD4 - sushi domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037175.2 | Intron | NP_001032252.1 | ||||
NM_017982.3 | Intron | NP_060452.3 | ||||
XM_005273169.1 | Intron | XP_005273226.1 | ||||
XM_005273172.1 | Intron | XP_005273229.1 | ||||
XM_006711408.1 | Intron | XP_006711471.1 | ||||
XM_011509685.1 | Intron | XP_011507987.1 | ||||
XM_011509687.1 | Intron | XP_011507989.1 | ||||
XM_017001583.1 | Intron | XP_016857072.1 | ||||
XM_017001584.1 | Intron | XP_016857073.1 | ||||
XM_017001585.1 | Intron | XP_016857074.1 | ||||
XM_017001586.1 | Intron | XP_016857075.1 | ||||
XM_017001587.1 | Intron | XP_016857076.1 | ||||
XM_017001588.1 | Intron | XP_016857077.1 | ||||
XM_017001589.1 | Intron | XP_016857078.1 |