Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120920 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD46 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD46 - CD46 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002389.4 | Intron | NP_002380.3 | ||||
NM_153826.3 | Intron | NP_722548.1 | ||||
NM_172350.2 | Intron | NP_758860.1 | ||||
NM_172351.2 | Intron | NP_758861.1 | ||||
NM_172352.2 | Intron | NP_758862.1 | ||||
NM_172353.2 | Intron | NP_758863.1 | ||||
NM_172359.2 | Intron | NP_758869.1 | ||||
NM_172361.2 | Intron | NP_758871.1 | ||||
XM_011509563.1 | Intron | XP_011507865.1 | ||||
XM_011509564.1 | Intron | XP_011507866.1 | ||||
XM_017001308.1 | Intron | XP_016856797.1 | ||||
XM_017001309.1 | Intron | XP_016856798.1 | ||||
XM_017001310.1 | Intron | XP_016856799.1 |