Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616027 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANLN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANLN - anillin actin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284301.2 | Intron | NP_001271230.1 | ||||
NM_001284302.2 | Intron | NP_001271231.1 | ||||
NM_018685.4 | Intron | NP_061155.2 | ||||
XM_006715746.1 | Intron | XP_006715809.1 | ||||
XM_006715747.3 | Intron | XP_006715810.1 | ||||
XM_017012354.1 | Intron | XP_016867843.1 | ||||
XM_017012355.1 | Intron | XP_016867844.1 | ||||
XM_017012356.1 | Intron | XP_016867845.1 |
KIAA0895 - KIAA0895 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |