Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156491 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MBTD1 - mbt domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017643.2 | 3768 | UTR 3 | NP_060113.2 | |||
XM_005257467.4 | 3768 | UTR 3 | XP_005257524.1 | |||
XM_005257468.4 | 3768 | Intron | XP_005257525.1 | |||
XM_011524922.2 | 3768 | UTR 3 | XP_011523224.1 | |||
XM_011524923.2 | 3768 | UTR 3 | XP_011523225.1 | |||
XM_011524924.2 | 3768 | UTR 3 | XP_011523226.1 | |||
XM_011524925.1 | 3768 | UTR 3 | XP_011523227.1 | |||
XM_011524926.2 | 3768 | UTR 3 | XP_011523228.1 | |||
XM_011524928.2 | 3768 | UTR 3 | XP_011523230.1 | |||
XM_011524929.2 | 3768 | UTR 3 | XP_011523231.1 | |||
XM_011524930.1 | 3768 | UTR 3 | XP_011523232.1 | |||
XM_011524931.2 | 3768 | UTR 3 | XP_011523233.1 | |||
XM_011524933.2 | 3768 | UTR 3 | XP_011523235.1 | |||
XM_011524934.2 | 3768 | Intron | XP_011523236.1 | |||
XM_011524935.2 | 3768 | Intron | XP_011523237.1 | |||
XM_011524936.1 | 3768 | Intron | XP_011523238.1 | |||
XM_017024782.1 | 3768 | UTR 3 | XP_016880271.1 |
NME1-NME2 - NME1-NME2 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME2 - NME/NM23 nucleoside diphosphate kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |