Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCCACAAAACCTAGTAGAAAA[C/G]TCACCCTCCAGACTGTGGCCAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA5A - carbonic anhydrase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001739.1 | Intron | NP_001730.1 | ||||
XM_005256134.4 | Intron | XP_005256191.1 | ||||
XM_011523309.2 | Intron | XP_011521611.1 | ||||
XM_011523310.2 | Intron | XP_011521612.1 | ||||
XM_017023646.1 | Intron | XP_016879135.1 |