Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGGCGAGCCTGTCTTCAGGTT[C/T]CCTAATGGTGTGTGCAGGTGCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OAS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OAS2 - 2'-5'-oligoadenylate synthetase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032731.1 | Intron | NP_001027903.1 | ||||
NM_002535.2 | Intron | NP_002526.2 | ||||
NM_016817.2 | Intron | NP_058197.2 |
Set Membership: |
HapMap |