Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGATAGTAAGTGCTATTTGGTAG[C/T]ACAATAGGGTGACTGTAGTCAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105370027 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105370027 - uncharacterized LOC105370027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM233 - transmembrane protein 233 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136534.1 | Intron | NP_001130006.1 | ||||
XM_005253880.3 | Intron | XP_005253937.1 | ||||
XM_005253881.3 | Intron | XP_005253938.1 | ||||
XM_011538328.2 | Intron | XP_011536630.1 | ||||
XM_011538329.2 | Intron | XP_011536631.1 | ||||
XM_011538330.2 | Intron | XP_011536632.1 | ||||
XM_011538331.2 | Intron | XP_011536633.1 |