Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601179 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAN - RAN, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300796.1 | 278 | UTR 5 | NP_001287725.1 | |||
NM_001300797.1 | 278 | UTR 5 | NP_001287726.1 | |||
NM_006325.4 | 278 | Silent Mutation | GTA,GTG | V,V 63 | NP_006316.1 | |
XM_017019772.1 | 278 | Silent Mutation | GTA,GTG | V,V 63 | XP_016875261.1 | |
XM_017019773.1 | 278 | UTR 5 | XP_016875262.1 |
Set Membership: |
HapMap |