Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116896 MIM: 605300 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CUX1 - cut like homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4285 - microRNA 4285 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2B2 - SH2B adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020979.4 | Intron | NP_066189.3 | ||||
XM_005276975.4 | Intron | XP_005277032.1 | ||||
XM_005276976.3 | Intron | XP_005277033.1 | ||||
XM_005276977.4 | Intron | XP_005277034.1 | ||||
XM_005276979.3 | Intron | XP_005277036.1 | ||||
XM_011515740.2 | Intron | XP_011514042.1 | ||||
XM_017011680.1 | Intron | XP_016867169.1 | ||||
XM_017011681.1 | Intron | XP_016867170.1 |
Set Membership: |
HapMap |