Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCACTCCCTCCGTCTCCCGTTC[A/G]ACAAACCCCAAACGCCTGTCTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601575 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ31356 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLJ31356 - uncharacterized protein FLJ31356 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOSL2 - FOS like 2, AP-1 transcription factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005253.3 | 1142 | Intron | NP_005244.1 | |||
XM_005264231.3 | 1142 | Intron | XP_005264288.1 | |||
XM_006711976.2 | 1142 | Intron | XP_006712039.1 | |||
XM_006711977.3 | 1142 | UTR 5 | XP_006712040.1 | |||
XM_017003737.1 | 1142 | UTR 5 | XP_016859226.1 |
Set Membership: |
HapMap |