Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAGGGCAAGGAGCTGCTCTGGTC[C/T]CAGCAGCCCTCCAGGCACCTGTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602635 MIM: 614988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEAF1 - DEAF1, transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPS8L2 - EPS8 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM80 - transmembrane protein 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042463.2 | Intron | NP_001035928.2 | ||||
NM_001276253.1 | Intron | NP_001263182.1 | ||||
NM_001276274.1 | Intron | NP_001263203.1 | ||||
NM_174940.3 | Intron | NP_777600.3 | ||||
XM_006718206.3 | Intron | XP_006718269.3 | ||||
XM_017017600.1 | Intron | XP_016873089.1 |