Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTTAAATTTAATAGTCATTTT[C/T]ATTACTGATTTTTATCGTTACAGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600629 MIM: 170261 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-DOB PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HLA-DOB - major histocompatibility complex, class II, DO beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAP2 - transporter 2, ATP binding cassette subfamily B member | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000544.3 | Intron | NP_000535.3 | ||||
NM_001290043.1 | Intron | NP_001276972.1 | ||||
NM_018833.2 | Intron | NP_061313.2 |
Set Membership: |
HapMap |