Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTAAAGCAGAGTGTGGTTCCC[A/C]TGTTAAAAAACAAAAGGAAACAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608994 MIM: 600772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKS1A - ankyrin repeat and sterile alpha motif domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015245.2 | Intron | NP_056060.2 | ||||
XM_005248964.3 | Intron | XP_005249021.1 | ||||
XM_006715036.2 | Intron | XP_006715099.1 | ||||
XM_011514431.2 | Intron | XP_011512733.1 | ||||
XM_011514432.2 | Intron | XP_011512734.1 | ||||
XM_011514433.1 | Intron | XP_011512735.1 | ||||
XM_011514434.2 | Intron | XP_011512736.1 | ||||
XM_011514435.2 | Intron | XP_011512737.1 | ||||
XM_011514436.2 | Intron | XP_011512738.1 | ||||
XM_011514437.2 | Intron | XP_011512739.1 | ||||
XM_017010593.1 | Intron | XP_016866082.1 |
TAF11 - TATA-box binding protein associated factor 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |