Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610257 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC31A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEC31A - SEC31 homolog A, COPII coat complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077206.3 | Intron | NP_001070674.1 | ||||
NM_001077207.3 | Intron | NP_001070675.1 | ||||
NM_001077208.3 | Intron | NP_001070676.1 | ||||
NM_001191049.2 | Intron | NP_001177978.1 | ||||
NM_001300744.2 | Intron | NP_001287673.1 | ||||
NM_001300745.2 | Intron | NP_001287674.1 | ||||
NM_001318119.1 | Intron | NP_001305048.1 | ||||
NM_001318120.1 | Intron | NP_001305049.1 | ||||
NM_016211.4 | Intron | NP_057295.2 |
Set Membership: |
HapMap |