Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604724 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPSE PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HPSE - heparanase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098540.2 | 1292 | Silent Mutation | AAC,AAT | N,N 459 | NP_001092010.1 | |
NM_001166498.2 | 1292 | Silent Mutation | AAC,AAT | N,N 385 | NP_001159970.1 | |
NM_001199830.1 | 1292 | Silent Mutation | AAC,AAT | N,N 401 | NP_001186759.1 | |
NM_006665.5 | 1292 | Silent Mutation | AAC,AAT | N,N 459 | NP_006656.2 |
Set Membership: |
HapMap |