Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCTCTGGCCCAGAGGCCTTC[C/T]CGTTGGCTCAGGTGGTCTCTCGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC442028 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC442028 - uncharacterized LOC442028 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEKT4 - tektin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286559.1 | Intron | NP_001273488.1 | ||||
NM_144705.3 | Intron | NP_653306.1 | ||||
XM_005263876.2 | Intron | XP_005263933.1 | ||||
XM_011510670.1 | Intron | XP_011508972.1 |