Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO3B - myosin IIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083615.3 | Intron | NP_001077084.2 | ||||
NM_138995.4 | Intron | NP_620482.3 | ||||
XM_006712299.3 | Intron | XP_006712362.1 | ||||
XM_011510654.2 | Intron | XP_011508956.1 | ||||
XM_011510655.2 | Intron | XP_011508957.1 | ||||
XM_011510656.2 | Intron | XP_011508958.1 | ||||
XM_011510657.2 | Intron | XP_011508959.1 | ||||
XM_011510658.2 | Intron | XP_011508960.1 |