Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608438 | ||||||||||||||||||||
Literature Links: |
TLK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TLK1 - tousled like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136554.1 | 2510 | Silent Mutation | GTA,GTG | V,V 595 | NP_001130026.1 | |
NM_001136555.1 | 2510 | Silent Mutation | GTA,GTG | V,V 547 | NP_001130027.1 | |
NM_012290.4 | 2510 | Silent Mutation | GTA,GTG | V,V 643 | NP_036422.3 | |
XM_011512238.2 | 2510 | Silent Mutation | GTA,GTG | V,V 580 | XP_011510540.1 | |
XM_017005421.1 | 2510 | Silent Mutation | GTA,GTG | V,V 426 | XP_016860910.1 | |
XM_017005422.1 | 2510 | Silent Mutation | GTA,GTG | V,V 426 | XP_016860911.1 |